Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

446726

Sigma-Aldrich

N-(Benzyloxycarbonyl)-2-aminoacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONHCH2CN
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

61-63 °C (lit.)

functional group

amine
nitrile
phenyl

SMILES string

O=C(NCC#N)OCc1ccccc1

InChI

1S/C10H10N2O2/c11-6-7-12-10(13)14-8-9-4-2-1-3-5-9/h1-5H,7-8H2,(H,12,13)

InChI key

DVUXKEFDAGQPQU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

446726-BULK:
446726-5G:4548173152431
446726-VAR:
446726-25G:4548173152424


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Versari et al.
Biotechnology and bioengineering, 79(1), 9-14 (2007-06-27)
A mutant of the cysteine protease papain, displaying nitrile hydratase and amidase activities, was expressed in Pichia pastoris and used for the hydrolysis of peptide nitriles in aqueous-organic media. The rate of hydrolysis of these nitriles is lowered by increasing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service