Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

446033

Sigma-Aldrich

Fmoc-2-Abz-OH

97%

Synonym(s):

2-(Fmoc-amino)benzoic acid, N-Fmoc-anthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17NO4
CAS Number:
Molecular Weight:
359.37
Beilstein:
7659726
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

216-218 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

SMILES string

OC(=O)c1ccccc1NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H17NO4/c24-21(25)18-11-5-6-12-20(18)23-22(26)27-13-19-16-9-3-1-7-14(16)15-8-2-4-10-17(15)19/h1-12,19H,13H2,(H,23,26)(H,24,25)

InChI key

CNAVPEPPAVHHKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

446033-VAR:
446033-1G:
446033-5G:
446033-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihai Ciustea et al.
Journal of medicinal chemistry, 51(20), 6563-6570 (2008-09-24)
Variola virus, the causative agent of smallpox, is a potential bioweapon. The development of new antiviral compounds for smallpox prophylaxis and treatment is critical, especially because the virus can acquire resistance to the drugs that are currently available. We have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service