Skip to Content
Merck
All Photos(1)

Documents

Safety Information

44161

Sigma-Aldrich

(2-Dodecen-1-yl)succinic anhydride

technical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.2 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

grade

technical

form

solid

bp

180-182 °C/5 mmHg (lit.)

mp

41-43 °C (lit.)
~45 °C

SMILES string

CCCCCCCCC\C=C/CC1CC(=O)OC1=O

InChI

1S/C16H26O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h10-11,14H,2-9,12-13H2,1H3/b11-10+

InChI key

UYCICMIUKYEYEU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(2-Dodecen-1-yl)succinic anhydride is a carboxylic acid anhydride. It participates in the synthesis of alkenyl inulin derivatives and biosurfactants.

Application

(2-Dodecen-1-yl)succinic anhydride may be used in the following:
  • Synthesis of amphiphilic N-[(2´-(dodec-2´-en-10-yl)succinoyl]chitosan.
  • Chemical modification of sago starch, via esterification.
  • Preparation of dodecenyl succinic anhydride (DDSA) gum arabic derivatives, via chemical modification of gum arabic.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

44161-250G:
44161-1KG:
44161-BULK:
44161-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and emulsification properties of dodecenyl succinic anhydride derivatives of gum Arabic.
Wang H, et al.
Food Hydrocolloids, 37, 143-148 (2014)
The effect of anhydride modification of sago starch on the tensile and water absorption properties of sago-filled linear low-density polyethylene (LLDPE).
Abdul Khalil HPS, et al.
Polymer-Plastics Technology and Engineering, 40(3), 249-263 (2001)
S Kokubun et al.
Biomacromolecules, 14(8), 2830-2836 (2013-06-26)
Biosurfactants have been synthesized using a low energy, environmentally friendly process by the derivatization of inulin with octenyl (OSA) and dodecenyl (DDSA) succinic anhydrides in aqueous solution. The inulin and its derivatives have been characterized using gel permeation chromatography/multi angle
Amphiphilic N-[2 (3)-(dodec-2'-en-1'-yl) succinoyl] chitosan: Synthesis and properties.
Tikhonov VE, et al.
Reactive functional Polymers, 68(2), 436-445 (2008)
Application of hydrophobic ?-cyclodextrin polymer in separation of metal ions by plasticized membranes.
Kozlowski CA, et al.
Separation and Purification Technology, 46(3), 136-144 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service