Skip to Content
Merck
All Photos(1)

Documents

Safety Information

441473

Sigma-Aldrich

tert-Butylperoxy isopropyl carbonate solution

75 wt. % in aliphatic hydrocarbons

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCO3C(CH3)3
CAS Number:
Molecular Weight:
176.21
UNSPSC Code:
12162002

concentration

75 wt. % in aliphatic hydrocarbons

refractive index

n20/D 1.448

density

0.87 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Org. Perox. C

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

441473-BULK:
441473-250ML:
441473-VAR:
441473-1L:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service