Skip to Content
Merck
All Photos(2)

Documents

Safety Information

425176

Sigma-Aldrich

Gadolinium(III) trifluoromethanesulfonate

98%

Synonym(s):

Tris(trifluoromethanesulfonato)gadolinium, Gadolinium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Gd
CAS Number:
Molecular Weight:
604.46
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: gadolinium
reagent type: catalyst

SMILES string

[Gd+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Gd/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

DYOBTPTUHDTANY-UHFFFAOYSA-K

Application

A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

425176-VAR:
425176-BULK:
425176-25G:
425176-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Evans et al.
Journal of the American Chemical Society, 125(34), 10162-10163 (2003-08-21)
A highly enantioselective, quinone Diels-Alder reaction catalyzed by chiral samarium and gadolinium pyridyl-bis(oxazoline) (pybox) complexes has been developed. The reaction scope has been extended to include three quinones and five dienes, all of which exclusively provide the expected endo product
Kobayashi, S.; Nagayama, S.
Journal of the American Chemical Society, 119, 10049-10049 (1997)
Yamanaka et al.
Organic letters, 2(2), 159-161 (2000-05-18)
[reaction: see text] Ytterbium trifluoromethanesulfonate [Yb(OTf)3] catalyzed the imino ene reaction of N-tosyl aldimine with alpha-methylstyrene to give a homoallylamine in moderate yield. Furthermore, addition of a catalytic amount of chlorotrimethylsilane (TMSCI) dramatically enhanced the imino ene reaction.
Tomoaki Hamada et al.
Journal of the American Chemical Society, 125(10), 2989-2996 (2003-03-06)
Catalytic asymmetric aldol reactions in aqueous media have been developed using Pr(OTf)(3) and chiral bis-pyridino-18-crown-6 1. In the asymmetric aldol reaction using rare earth metal triflates (RE(OTf)(3)) and 1, slight changes in the ionic diameters of the metal cations greatly
The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service