Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

424587

Sigma-Aldrich

(4-Chlorobenzyl)triphenylphosphonium chloride

98%

Synonym(s):

[(4-Chlorophenyl)methyl]triphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2P(C6H5)3Cl
CAS Number:
Molecular Weight:
423.31
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

>300 °C (lit.)

functional group

chloro
phosphine

SMILES string

[Cl-].Clc1ccc(C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)cc1

InChI

1S/C25H21ClP.ClH/c26-22-18-16-21(17-19-22)20-27(23-10-4-1-5-11-23,24-12-6-2-7-13-24)25-14-8-3-9-15-25;/h1-19H,20H2;1H/q+1;/p-1

InChI key

RAHOAHBOOHXRDY-UHFFFAOYSA-M

Application

Reactant for the synthesis of:
  • β-amyloid plaque ligands
  • Selective norbornane-type aspartate analog inhibitors of glutamate transporter 1
  • Pyrethroids with insecticidal activity
  • Chlorinated aromatic avenaciolide analogs with antifungal activity
  • Arylethenylbenzofuroxan derivatives via Wittig-Boden reactions
  • Substituted benzofuroxans for inhibition of Trypanosoma cruzi growth
  • Styrylchromones via Wittig reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

424587-VAR:
424587-BULK:
424587-5G:
424587-25G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service