Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

424277

Sigma-Aldrich

Poly(isobutylene-co-maleic acid) sodium salt

cross-linked

Synonym(s):

Isobutylene-maleic anhydride copolymer sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H8)m(C4H4O4)n · xNa
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

crosslinking

cross-linked

particle size

24-40 μm

capacity

300 g/g absorption capacity (distilled water)
65 g/g absorption capacity (0.9 wt. % saline solution)

mp

>300 °C

density

1.3 g/mL at 25 °C (lit.)

SMILES string

CC(C)=C.OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(=O)O[Na]

Looking for similar products? Visit Product Comparison Guide

Application

Superabsorbent fiber. Amount of aqueous fluid absorbed decreases with increasing salt content. Stable up to 250°C

Physical form

Water-absorbing fiber,

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

424277-1KG:
424277-250G:
424277-BULK:
424277-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service