41762
1,9-Dimethylxanthine
≥98.0%
Synonym(s):
2,6-Dihydroxy-1,9-dimethylpurine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
Beilstein:
180792
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥98.0%
mp
≥300 °C
SMILES string
CN1C(=O)Nc2c(ncn2C)C1=O
Gene Information
rat ... Adora1(29290) , Adora2a(25369)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service