Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

415871

Sigma-Aldrich

Trimethylolpropane ethoxylate (1 EO/OH) methyl ether diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2CH2OCH2)2C(C2H5)CH2OCH2CH2OCH3
CAS Number:
Molecular Weight:
388.45
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

250 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.462 (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

CCC(COCCOC)(COCCOC(=O)C=C)COCCOC(=O)C=C

InChI

1S/C19H32O8/c1-5-17(20)26-12-10-24-15-19(7-3,14-23-9-8-22-4)16-25-11-13-27-18(21)6-2/h5-6H,1-2,7-16H2,3-4H3

InChI key

SBOXDRICSJHEBA-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

415871-1L:
415871-BULK:
415871-250ML:
415871-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service