Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

414646

Sigma-Aldrich

Boc-D-Ala-OMe

99%

Synonym(s):

N-(tert-Butoxycarbonyl)-D-alanine methyl ester, Boc-D-alanine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
203.24
Beilstein:
4310313
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D +45°, c = 1 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

34-37 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H](C)NC(=O)OC(C)(C)C

InChI

1S/C9H17NO4/c1-6(7(11)13-5)10-8(12)14-9(2,3)4/h6H,1-5H3,(H,10,12)/t6-/m1/s1

InChI key

GJDICGOCZGRDFM-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

414646-VAR:
414646-10G:
414646-BULK:
414646-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service