Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

414387

Sigma-Aldrich

Vanadyl 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine

Dye content 98 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H32N8O5V
CAS Number:
Molecular Weight:
947.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 98%

λmax

710 nm

SMILES string

O=[V]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7cc(Oc8ccccc8)ccc37)c9ccc(Oc%10ccccc%10)cc69)c%11ccc(Oc%12ccccc%12)cc5%11)c%13ccc(Oc%14ccccc%14)cc4%13

InChI

1S/C56H32N8O4.O.V/c1-5-13-33(14-6-1)65-37-21-25-41-45(29-37)53-57-49(41)62-54-47-31-39(67-35-17-9-3-10-18-35)23-27-43(47)51(59-54)64-56-48-32-40(68-36-19-11-4-12-20-36)24-28-44(48)52(60-56)63-55-46-30-38(66-34-15-7-2-8-16-34)22-26-42(46)50(58-55)61-53;;/h1-32H;;/q-2;;+2

InChI key

NGSMDHYQABJENH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

414387-BULK:
414387-VAR:
414387-1G:
414387-250MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdullah Haaziq Ahmad Makinudin et al.
Nanoscale research letters, 10, 53-53 (2015-04-09)
The use of templating method to synthesize the vanadyl 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine (VOPcPhO):[6,6]-phenyl C71 butyric acid methyl ester (PC71BM) composite nanotubes is presented here. VOPcPhO is a p-type material and PC71BM is an n-type material which acts as an electron donor and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service