Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

412600

Sigma-Aldrich

4-tert-Butyl-2-nitrophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(NO2)OH
CAS Number:
Molecular Weight:
195.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

97 °C/1 mmHg (lit.)

mp

27-29 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

CC(C)(C)c1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C10H13NO3/c1-10(2,3)7-4-5-9(12)8(6-7)11(13)14/h4-6,12H,1-3H3

InChI key

IHGNADPMUSNTJW-UHFFFAOYSA-N

Application

4-tert-Butyl-2-nitrophenol may be used for the preparation of (2-hydroxy-3-nitro-5-tert-butylbenzyl)trimethylammonium iodide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

412600-VAR:
412600-25G:
412600-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensations of N-arylhydroxylamines for the preparation of 5,5'-di-tert-butyl-2,2'-dihydroxydiphenylamine.
Spence JD, et al.
Tetrahedron Letters, 44(4), 849-851 (2003)
Vitaly A Osyanin et al.
The Journal of organic chemistry, 79(3), 1192-1198 (2014-01-15)
The first example of the use of potassium trinitromethanide as a 1,1-ambiphilic synthon equivalent for the construction of a benzofuran moiety mediated by triethylamine has been developed. The method tolerates a variety of functional groups on the starting quaternary ammonium
Yong Zhang et al.
Analytica chimica acta, 899, 75-84 (2015-11-10)
In this research, the applicability of a new sorbent based on boronate affinity material is demonstrated. For this purpose, six strong polar nitrophenols were selected as models which are difficult to be extracted in neutral form (only based on hydrophobic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service