Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

411655

Sigma-Aldrich

Allyl cyclohexanepropionate

98%

Synonym(s):

2-Propenyl 3-cyclohexylpropanoate, Allyl 3-cyclohexylpropionate, Cyclohexanepropanoic acid 2-propen-1-yl ester, Cyclohexanepropionic acid allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.46 (lit.)

bp

91 °C/1 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

C=CCOC(=O)CCC1CCCCC1

InChI

1S/C12H20O2/c1-2-10-14-12(13)9-8-11-6-4-3-5-7-11/h2,11H,1,3-10H2

InChI key

TWXUTZNBHUWMKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

411655-VAR:
411655-BULK:
411655-250ML:
411655-50ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service