Skip to Content
Merck
All Photos(1)

Key Documents

411620

Sigma-Aldrich

Hydrogen hexachloroiridate(IV) hydrate

4 wt. % in H2O

Synonym(s):

Dihydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Hydrogen iridium hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2Cl6Ir · xH2O
CAS Number:
Molecular Weight:
406.95 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302

form

liquid

concentration

4 wt. % in H2O

impurities

2-2.2% Ir

mp

65 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].[H]O[H].Cl[Ir](Cl)(Cl)Cl

InChI

1S/6ClH.Ir.H2O/h6*1H;;1H2/q;;;;;;+4;/p-4

InChI key

YDVQBPXDKJKDME-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Catalyzes the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes. Utilized in the formation of iridium-substituted Dawson- and Keggin-type polyoxometallates by refilling the vacant site of the lacunary precursors.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service