Skip to Content
Merck
All Photos(1)

Key Documents

410160

Sigma-Aldrich

Iron(II) phthalocyanine bis(pyridine) complex

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H26FeN10
CAS Number:
Molecular Weight:
726.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 95%

λmax

655 nm

SMILES string

c1ccncc1.c2ccncc2.[Fe]3n4c5nc6nc(nc7n3c(nc8nc(nc4c9ccccc59)c%10ccccc8%10)c%11ccccc7%11)c%12ccccc6%12


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service