408883
Nickel(II) 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine
Dye content 97 %
Synonym(s):
1,4,8,11,15,18,22,25-Octabutoxyphthalocyanine nickel(II) salt
About This Item
Recommended Products
composition
Dye content, 97%
reaction suitability
reagent type: catalyst
core: nickel
mp
288-293 °C (lit.)
λmax
743 nm
SMILES string
CCCCOc1ccc(OCCCC)c2c3nc(nc4n5[Ni]n6c(n3)c7c(OCCCC)ccc(OCCCC)c7c6nc8nc(nc5c9c(OCCCC)ccc(OCCCC)c49)c%10c(OCCCC)ccc(OCCCC)c8%10)c12
InChI
1S/C64H80N8O8.Ni/c1-9-17-33-73-41-25-26-42(74-34-18-10-2)50-49(41)57-65-58(50)70-60-53-45(77-37-21-13-5)29-30-46(78-38-22-14-6)54(53)62(67-60)72-64-56-48(80-40-24-16-8)32-31-47(79-39-23-15-7)55(56)63(68-64)71-61-52-44(76-36-20-12-4)28-27-43(75-35-19-11-3)51(52)59(66-61)69-57;/h25-32H,9-24,33-40H2,1-8H3;/q-2;+2
InChI key
JFVRZEMMDHQJPH-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PRTR
Specified Class I Designated Chemical Substances
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
408883-VAR:
408883-500MG:4548173918280
408883-100MG:
408883-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service