Skip to Content
Merck
All Photos(1)

Key Documents

405159

Sigma-Aldrich

IR-1135

Dye content 95 %

Synonym(s):

8-[[3-[(6,7-Dihydro-2,4-diphenyl-5H-1-benzopyran-8-yl)methylene]-2-phenyl-1-cyclopenten-1-yl]methylene]-5,6,7,8-tetrahydro-2,4-diphenyl-1-benzopyrylium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H45BF4O2
CAS Number:
Molecular Weight:
824.75
UNSPSC Code:
12171500

composition

Dye content, 95%

SMILES string

F[B-](F)(F)F.C1CC(\C=C2/CCC(\C=C3/CCCc4c(cc([o+]c34)-c5ccccc5)-c6ccccc6)=C2c7ccccc7)=C8OC(=CC(=C8C1)c9ccccc9)c%10ccccc%10

Application

Q-switching laser dye

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service