40291
Ammonium hydroxide solution
≥25% NH3 in H2O, semiconductor grade VLSI PURANAL™ (Honeywell 17605)
Synonym(s):
Ammonia aqueous, Ammonia water
About This Item
Recommended Products
grade
semiconductor grade VLSI PURANAL™ (Honeywell 17605)
form
liquid
CofA
specification on request
expl. lim.
27 %
concentration
≥25% NH3 in H2O
density
0.9 g/mL at 25 °C (lit.)
SMILES string
[NH4+].[OH-]
InChI
1S/H3N.H2O/h1H3;1H2
InChI key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
- precipitant for the R2O3 group of elements,
- complexing agent,
- dissolution medium.7
Application
- to clean wafers.1
- to prepare magnetic iron oxide nanoparticles.
- in the recovery of cellulose from sugarcane bagasse. The process enhances hydrolysis and production of bioethanol.
- in bi-directional perfusion technique of human periodontal ligament cell sheets.
- as a mobile phase in the liquid chromatography for prebiotic galactooligosaccharides.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
40291-2.5L:
40291-VAR:
40291-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service