401811
Sodium pentamethylcyclopentadienide solution
0.5 M in THF
Synonym(s):
(Pentamethylcyclopentadienyl)sodium, Sodium pentamethylcyclopentadiene
About This Item
Recommended Products
form
liquid
reaction suitability
reagent type: ligand
concentration
0.5 M in THF
density
0.905 g/mL at 25 °C
SMILES string
CC1=C(C)C(C)([Na])C(C)=C1C
InChI
1S/C10H15.Na/c1-6-7(2)9(4)10(5)8(6)3;/h1-5H3;
InChI key
NWRNNTRBUPLTKV-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PRTR
Class I Designated Chemical Substances
FSL
Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
401811-VAR:
401811-BULK:
401811-50ML:4548173145051
401811-100ML:4548173145044
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service