Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

399728

Sigma-Aldrich

(R)-(−)-N-[1-(1-Naphthyl)ethyl]phthalamic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[C10H7CH(CH3)NHCO]C6H4CO2H
CAS Number:
Molecular Weight:
319.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]26/D −48°, c = 1 in methanol

mp

166 °C (dec.) (lit.)

functional group

amide
carboxylic acid

SMILES string

C[C@@H](NC(=O)c1ccccc1C(O)=O)c2cccc3ccccc23

InChI

1S/C20H17NO3/c1-13(15-12-6-8-14-7-2-3-9-16(14)15)21-19(22)17-10-4-5-11-18(17)20(23)24/h2-13H,1H3,(H,21,22)(H,23,24)/t13-/m1/s1

InChI key

ZEKKAEAOTSMLNW-CYBMUJFWSA-N

General description

N-[1-(1-Naphthyl)ethyl]phthalamic acid is a chiral nonsteroidal anti-inflammatory drug.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

399728-1G:
399728-BULK:
399728-250MG:
399728-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonaqueous capillary electrophoresis chiral separations with quaternary ammonium β-cyclodextrin
Wang F and Khaledi MG
Journal of Chromatography A, 817(1-2), 121-128 (1998)
Yu-Tong Jiang et al.
The New phytologist, 225(4), 1606-1617 (2019-10-01)
Two types of tonoplast proton pumps, H+ -pyrophosphatase (V-PPase) and the H+ -ATPase (V-ATPase), establish the proton gradient that powers molecular traffic across the tonoplast thereby facilitating turgor regulation and nutrient homeostasis. However, how proton pumps regulate development remains unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service