Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

388564

Sigma-Aldrich

Sodium borodeuteride

96 atom % D

Synonym(s):

Sodium tetradeuterioboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBD4
CAS Number:
Molecular Weight:
41.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

96 atom % D

Quality Level

autoignition temp.

752 °F

density

1.184 g/mL

mass shift

M+4

SMILES string

[Na+].[2H][B-]([2H])([2H])[2H]

InChI

1S/BH4.Na/h1H4;/q-1;+1/i1D4;

InChI key

YOQDYZUWIQVZSF-XWFVQAFUSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in screw-cap bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 3: Spontaneously combustible substances and water- reactive materials
Metal hydrides
Hazardous rank I
1st spontaneously combustible materials and water reactive materials

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

388564-VAR:
388564-BULK:
388564-5G:
388564-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khalef Lefsih et al.
Phytochemistry, 154, 47-55 (2018-07-15)
This work aims to fill the gap in the present knowledge about the structure of pectin from Opuntia ficus-indica. The water-soluble pectin (WSP) fraction, extracted with the Microwave Assisted Extraction (MAE), was further deproteinated (dWSP) and analyzed through several biophysical
Utoomporn Surayot et al.
Bioscience, biotechnology, and biochemistry, 79(10), 1644-1653 (2015-05-15)
Sulfated polysaccharides (SP) isolated from freshwater green algae, Spirogyra neglecta (Hassall) Kützing, and fractionated SPs were examined to investigate their molecular characteristics and immunomodulatory activity. The crude and fractionated SPs (F1, F2, and F3) consisted mostly of carbohydrates (68.5-85.3%), uronic
Maliha Shaikh et al.
Clinica chimica acta; international journal of clinical chemistry, 442, 24-32 (2015-01-17)
Measurement of intestinal permeability is important in several diseases but currently several methods are employed. We sought to: (1) develop a new GC based method to measure urinary mannitol, lactulose and sucralose to assess regional and total gut permeability; (2)
Guoyun Li et al.
Journal of chromatography. A, 1397, 43-51 (2015-04-29)
Pneumococcal type-3 polysaccharide (Pn3P) is considered a major target for the development of a human vaccine to protect against Streptococcus pneumoniae infection. Thus, it is critical to develop methods for the preparation and analysis of Pn3P-derived oligosaccharides to better understand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service