Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

38221

Sigma-Aldrich

Diisooctyldithiophosphinic acid

technical, ~85% (T)

Synonym(s):

Bis(2,4,4-trimethylpentyl)dithiophosphinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H35PS2
CAS Number:
Molecular Weight:
322.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

~85% (T)

refractive index

n20/D 1.507

density

0.95 g/mL at 20 °C (lit.)

SMILES string

CC(CC(C)(C)C)CP(S)(=S)CC(C)CC(C)(C)C

InChI

1S/C16H35PS2/c1-13(9-15(3,4)5)11-17(18,19)12-14(2)10-16(6,7)8/h13-14H,9-12H2,1-8H3,(H,18,19)

InChI key

YOCZZJWFWDUAAR-UHFFFAOYSA-N

General description

Diisooctyldithiophosphinic acid has been reported as soft base. As per Pearson′s Hard and Soft Acids and Bases principle since soft base has an affinity for soft acid, it exhibits high affinity for soft cations.[1]

Application

Complexing agent for the extraction of transition metals
Diisooctyldithiophosphinic acid (bis(2,4,4-trimethyl pentyl)dithiophosphinic acid, cyanex 301) may be used in the following studies:
  • As mobile carrier in the preparation of new supported liquid membrane (SLM) system, useful for the selective transport of bismuth ions.[2]
  • As an efficient carrier for the preparation of bulk liquid membrane (BLM) system, utilized for the transport of methylene blue.[3]
  • As carrier in the preparation of a novel bulk liquid membrane (BLM) system, used to evaluate mercury transport and recovery of the ions from acidic nitrate solutions.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

38221-500ML:
38221-VAR:
38221-25ML:
38221-BULK:
38221-100ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient and selective transport of methylene blue through a bulk liquid membrane containing Cyanex 301 as carrier.
Madaeni SS, et al.
Separation and Purification Technology, 81(2), 116-123 (2011)
Highly efficient and selective transport of Hg2+ ions through a bulk liquid membrane containing Cyanex 301 as carrier.
Madaeni SS, et al.
Asia-Pacific Journal of Chemical Engineering, 6(4), 631-638 (2011)
Effects of membrane characteristics on selective transport of bismuth ion through supported liquid membrane
Madaeni SS and Karami Zand HR.
9th Iranian Chemical Engineering Congress (IChEC9), Iran University of Science and Technology (IUST, 23, 25-25 (2004)
Selective transport of bismuth ions through supported liquid membrane.
Madaeni SS and Karami Zand HR.
Chemical Engineering & Technology, 28(8), 892-892 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service