Skip to Content
Merck
All Photos(1)

Documents

Safety Information

380652

Sigma-Aldrich

3,4-O-Isopropylidene-L-threonic acid calcium salt

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22CaO10
CAS Number:
Molecular Weight:
390.40
Beilstein:
5670587
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +24°, c = 1 in H2O

mp

>300 °C (lit.)

SMILES string

CC1(C)OC[C@@H](O1)[C@H](O)C(=O)O[Ca]OC(=O)[C@@H](O)[C@H]2COC(C)(C)O2

InChI

1S/2C7H12O5.Ca/c2*1-7(2)11-3-4(12-7)5(8)6(9)10;/h2*4-5,8H,3H2,1-2H3,(H,9,10);/q;;+2/p-2/t2*4-,5+;/m11./s1

InChI key

NOUVZISPTGHJFE-ASZNEFJQSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

380652-VAR:
380652-BULK:
380652-1G:
380652-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service