Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

379182

Sigma-Aldrich

m-Carborane

98%

Synonym(s):

1,7-Dicarbadodecaborane(12)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H12B10
CAS Number:
Molecular Weight:
144.23
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

storage temp.

2-8°C

SMILES string

B1BBBBC2BC(BBB1)B2

InChI

1S/C2H12B10/c3-1-4-2(3)6-8-10-12-11-9-7-5-1/h1-12H

InChI key

VNANXKFVYVNBPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

379182-500MG:
379182-1G:
379182-BULK:
379182-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service