Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

368709

Sigma-Aldrich

2-Chloro-5-fluoronitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)NO2
CAS Number:
Molecular Weight:
175.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

238 °C (lit.)

mp

37-40 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(F)ccc1Cl

InChI

1S/C6H3ClFNO2/c7-5-2-1-4(8)3-6(5)9(10)11/h1-3H

InChI key

DVXDJQKEEKXJBW-UHFFFAOYSA-N

General description

2-Chloro-5-fluoronitrobenzene is a fluorinated building block.

Application

2-Chloro-5-fluoronitrobenzene may be used in the preparation of N,N-dimethyl-2-(2-amino-4-fluorophenylthio)benzylamine. It may be used in the synthesis of N-methyl-2-(2-amino-4-fluorophenylthio)benzylamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

368709-1G:
368709-BULK:
368709-5G:
368709-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS AND EVALUATION OF N-METHYL-2-(2-AMINO-4-[18 F] FLUOROPHENYLTHIO) BENZYLAMINE AS SERT IMAGING AGENT.
Huang Y, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 52 (2009)
Grace G Shiue et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 44(12), 1890-1897 (2003-12-09)
There has been considerable interest in the development of PET radioligands that are useful for imaging serotonin transporter (SERT) in the living human brain. For the last decade, (11)C-(+)McN5652 has been the most promising PET agent for studying SERT in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service