Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

367826

Sigma-Aldrich

1-(p-Toluenesulfonyl)azetidine

97%

Synonym(s):

1-Tosylazetidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2S
CAS Number:
Molecular Weight:
211.28
Beilstein:
177046
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

119-122 °C (lit.)

functional group

sulfonamide

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2CCC2

InChI

1S/C10H13NO2S/c1-9-3-5-10(6-4-9)14(12,13)11-7-2-8-11/h3-6H,2,7-8H2,1H3

InChI key

VKCBXONEZGIOSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(p-Toluenesulfonyl)azetidine is a heterocyclic building block. Ag(I)-catalyzed ring-opening of 1-(p-toluenesulfonyl)azetidine (N-tosylazetidine) with alcohols, amines, thiols and related tethered 1,2-ethane dinucleophiles has been reported.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

367826-VAR:
367826-BULK:
367826-1G:
367826-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milan Bera et al.
The Journal of organic chemistry, 76(5), 1475-1478 (2011-02-05)
[Ag(COD)(2)]PF(6) catalyzes the ring-opening of N-tosylaziridines and -azetidines with alcohols, amines, thiols, and related tethered 1,2-ethane dinucleophiles. Initial rate studies and DFT-based evaluation of stepwise energetics suggest an inverse relationship between the nucleophilic reactivity of a heteroatom donor and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service