Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

362816

Sigma-Aldrich

4-(Nonyloxy)benzoic acid

97%

Synonym(s):

4-n -Nonyloxybenzoic acid, p -(Nonyloxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8OC6H4CO2H
CAS Number:
Molecular Weight:
264.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid crystal

SMILES string

CCCCCCCCCOc1ccc(cc1)C(O)=O

InChI

1S/C16H24O3/c1-2-3-4-5-6-7-8-13-19-15-11-9-14(10-12-15)16(17)18/h9-12H,2-8,13H2,1H3,(H,17,18)

InChI key

BOZLUAUKDKKZHJ-UHFFFAOYSA-N

Application

Intermediate for the synthesis of liquid crystals including chiral ferroelectric benzoates. Used in the preparation of medicinal compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

362816-VAR:
362816-BULK:
362816-10G:
362816-50G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 23-23 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service