Skip to Content
Merck
All Photos(1)

Key Documents

356417

Supelco

Niobium ICP/DCP standard solution

analytical standard, 10,000 μg/mL Nb in H2O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

contains

hydrofluoric acid as stabilizer (trace)

concentration

10,000 μg/mL Nb in H2O

technique(s)

AAS: suitable
ICP: suitable

format

single component solution


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service