Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

31546

Sigma-Aldrich

N,N-Diacetyl-O-methylhydroxylamine

≥95.0%

Synonym(s):

N-Acetyl-N-methoxyacetamide, N-Methoxydiacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
3537117
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

Assay

≥95.0%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.447

density

1.12 g/mL at 20 °C (lit.)

SMILES string

CON(C(C)=O)C(C)=O

InChI

1S/C5H9NO3/c1-4(7)6(9-3)5(2)8/h1-3H3

InChI key

HWZKQIRJZPOEQF-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of benzamide derivatives as rally available antitumor agents
  • Synthesis of hydroxyethylamine (HEA) BACE-1 inhibitors
  • Acetylation of amines (acetylating agent)

Other Notes

Chemoselective acetylating agent for prim. amines in the presence of sec. amines and alcohols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

31546-BULK:
31546-5ML:
31546-VAR:
31546-25ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Kikigawa et al.
Tetrahedron Letters, 31, 243-243 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service