Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

311391

Sigma-Aldrich

Barium phosphate dibasic

97%

Synonym(s):

Barium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaHPO4
CAS Number:
Molecular Weight:
233.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

97%

form

powder

SMILES string

[Ba++].OP([O-])([O-])=O

InChI

1S/Ba.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+2;/p-2

InChI key

LYSTYSFIGYAXTG-UHFFFAOYSA-L

Application

Reactant involved in hydrothermal synthesis of α-Ba2P2O7

Reactant involved in synthesis of modified carbon paste electrode for determination of cadmium and lead via differential pulse anodic stripping voltammetry

Involved in studies of the temperature dependence of soft mode frequency, dielectric constant and loss tangent of crystals in an electric field

Reagent used to asses glucose-specific poly(allylamine) hydrogels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

311391-250G:
311391-50G:4548173133324
311391-VAR:
311391-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orlando Laitano et al.
Frontiers in physiology, 9, 1496-1496 (2018-11-16)
Hyperthermia and dehydration can occur during exercise in hot environments. Nevertheless, whether elevations in extracellular osmolality contributes to the increased skeletal muscle tension, sarcolemmal injury, and oxidative stress reported in warm climates remains unknown. We simulated osmotic and heat stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service