Skip to Content
Merck
All Photos(1)

Key Documents

308331

Sigma-Aldrich

2′,3′-Dideoxyadenosine

97%

Synonym(s):

ddA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
Beilstein:
619924
EC Number:
MDL number:
UNSPSC Code:
12352103

Pricing and availability is not currently available.

Assay

97%

optical activity

[α]20/D −27°, c = 1 in H2O

mp

181-184 °C (lit.)

SMILES string

Nc1ncnc2n(cnc12)[C@H]3CC[C@@H](CO)O3

InChI

1S/C10H13N5O2/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(3-16)17-7/h4-7,16H,1-3H2,(H2,11,12,13)/t6-,7+/m0/s1

InChI key

WVXRAFOPTSTNLL-NKWVEPMBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Research tool for antiviral and anticancer studies.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 20, 52-52 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service