Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

303321

Sigma-Aldrich

Dimethylthexylsilyl trifluoromethanesulfonate

97%

Synonym(s):

Dimethyl-(2,3-dimethyl-2-butyl)silyl trifluoromethanesulfonate, TDS triflate, Thexyl-dimethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid thexyl-dimethylsilyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19F3O3SSi
CAS Number:
Molecular Weight:
292.39
Beilstein:
4186137
MDL number:
UNSPSC Code:
12352000

Assay

97%

bp

53-54 °C/0.6 mmHg (lit.)

density

1.16 g/mL at 20 °C (lit.)

SMILES string

CC(C)C(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

Protecting reagent for alcohols and other functional groups.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

303321-VAR:
303321-BULK:
303321-25G:
303321-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 19, 52-52 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service