Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

29514

Sigma-Aldrich

(R)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis(di-2-furylphosphine)

≥97%, optical purity ee: ≥99%

Synonym(s):

(R)-(+)-2,2′-Bis(di-2-furylphosphino)-6,6′-dimethoxy-1,1′-biphenyl, (R)-2-Furyl-MeOBIPHEP, SL-A108-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24O6P2
CAS Number:
Molecular Weight:
542.46
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97%

optical purity

ee: ≥99%

SMILES string

COc1cccc(P(c2ccco2)c3ccco3)c1-c4c(OC)cccc4P(c5ccco5)c6ccco6

InChI

1S/C30H24O6P2/c1-31-21-9-3-11-23(37(25-13-5-17-33-25)26-14-6-18-34-26)29(21)30-22(32-2)10-4-12-24(30)38(27-15-7-19-35-27)28-16-8-20-36-28/h3-20H,1-2H3

InChI key

ABGYJVGTLXSMBM-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

29514-1G:
29514-5G:
29514-BULK:
29514-100MG:
29514-VAR:
29514-500MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service