Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

290742

Sigma-Aldrich

4′-Phenoxyacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4COCH3
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

bp

200 °C/12 mmHg (lit.)

mp

50-52 °C (lit.)

SMILES string

CC(=O)c1ccc(Oc2ccccc2)cc1

InChI

1S/C14H12O2/c1-11(15)12-7-9-14(10-8-12)16-13-5-3-2-4-6-13/h2-10H,1H3

InChI key

DJNIFZYQFLFGDT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

290742-5G:
290742-25G:
290742-BULK:
290742-VAR:
290742-100G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service