Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28270

Sigma-Aldrich

Curing Agent C

≥95.0% (NT)

Synonym(s):

p-Toluenesulfonic acid morpholine salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3S · C4H9NO
CAS Number:
Molecular Weight:
259.32
Beilstein:
4042738
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Assay

≥95.0% (NT)

mp

123-126 °C

SMILES string

C1COCCN1.Cc2ccc(cc2)S(O)(=O)=O

InChI

1S/C7H8O3S.C4H9NO/c1-6-2-4-7(5-3-6)11(8,9)10;1-3-6-4-2-5-1/h2-5H,1H3,(H,8,9,10);5H,1-4H2

InChI key

LYWVNPSVLAFTFX-UHFFFAOYSA-N

Other Notes

Effective additive to reduce checking during hardening of paint films.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

28270-500G:
28270-BULK:
28270-VAR:
28270-INTR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Swift et al.
Offic. Dig., Federation Paint & VarnishProd. Clubs, 29, 664-664 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service