Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

276774

Sigma-Aldrich

1,2,3-Triphenylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3C6H3
CAS Number:
Molecular Weight:
306.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

157-159 °C (lit.)

SMILES string

c1ccc(cc1)-c2cccc(-c3ccccc3)c2-c4ccccc4

InChI

1S/C24H18/c1-4-11-19(12-5-1)22-17-10-18-23(20-13-6-2-7-14-20)24(22)21-15-8-3-9-16-21/h1-18H

InChI key

CHBDXRNMDNRJJC-UHFFFAOYSA-N

Application

1,2,3-Triphenylbenzene has been used in the synthesis of 1,2,3-tricyclohexylcyclohexane via catalytic hydrogenation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

276774-1G:
276774-BULK:
276774-5G:
276774-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sesha Bamini Nariyangadu et al.
ACS omega, 5(33), 20863-20873 (2020-09-03)
The synthesis and crystal structure of rhodamine 590 acid phthalate (RhAP) have been reported. This novel solid-state rhodamine derivative not only has a longer fluorescence lifetime compared to rhodamine solid-state matrixes where emission is quenched but also possesses strong nonlinear
Stereochemistry and Conformational Anomalies of 1, 2, 3-and 1, 2, 3, 4-Polycyclohexylcyclohexanes.
Columbus I, et al.
Journal of the American Chemical Society, 118(29), 6890-6896 (1996)
Enaam Jamal Al Dine et al.
Environmental science and pollution research international, 22(22), 17850-17859 (2015-07-15)
In this study, pine needles were used as cost-effective and reliable passive bio-monitors to concomitantly evaluate atmospheric concentrations of three classes of persistent organic pollutants, polychlorinated biphenyls (PCBs), organochlorine pesticides (OCPs), and polycyclic aromatic hydrocarbons (PAHs). The extraction of persistent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service