Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

262323

Sigma-Aldrich

Borane 4-methylmorpholine complex

97%

Synonym(s):

4-Methylmorpholine-borane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H14BNO
CAS Number:
Molecular Weight:
114.98
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

42-45 °C (lit.)

functional group

ether

SMILES string

[BH3-][O+]1CCN(C)CC1

InChI

1S/C5H14BNO/c1-7-2-4-8(6)5-3-7/h2-5H2,1,6H3

InChI key

LCJPHDKSLJEBCF-UHFFFAOYSA-N

Application

Reagent for:
  • Labeling oligosaccharides for quantitative mass spectrometry
  • Analysis of sugar markers associated with Bacillus anthracis spores after culture
  • Synthesis of sulfated red seaweed galactan derived disaccharide alditol
  • Analysis of sulfated galactans from red algae by reductive hydrolysis and mild methanolysis by HPLC

Reactant for methanolic cleavage

Used in reductive pyrolysis of carrageenan by high performance anion-exchange chromatography

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

262323-VAR:
262323-25G:
262323-BULK:
262323-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service