Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

256331

Sigma-Aldrich

(R)-(+)-1-Phenyl-1-propanol

99%

Synonym(s):

(R)-(+)-α-Ethylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(C6H5)OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
2041555
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D +48°, c = 2.25 in hexane

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.5196 (lit.)

bp

218-220 °C (lit.)

density

0.993 g/mL at 20 °C (lit.)

SMILES string

CC[C@@H](O)c1ccccc1

InChI

1S/C9H12O/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3/t9-/m1/s1

InChI key

DYUQAZSOFZSPHD-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

256331-100MG:
256331-VAR:
256331-BULK:
256331-5ML:
256331-1ML:
256331-500MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service