Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

256293

Sigma-Aldrich

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol

99%

Synonym(s):

(S)-(+)-α-Tetralol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
Beilstein:
2208779
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]17/D +32°, c = 2.5 in chloroform

optical purity

ee: 99% (HPLC)

bp

92 °C/1.8 mmHg (lit.)

mp

39-40 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

O[C@H]1CCCc2ccccc12

InChI

1S/C10H12O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10-11H,3,5,7H2/t10-/m0/s1

InChI key

JAAJQSRLGAYGKZ-JTQLQIEISA-N

Gene Information

General description

Chiral building block.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

256293-500MG:
256293-VAR:
256293-100MG:
256293-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service