Skip to Content
Merck
All Photos(1)

Documents

Safety Information

253766

Sigma-Aldrich

3-Chlorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NCS
CAS Number:
Molecular Weight:
169.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.6585 (lit.)

bp

249-250 °C (lit.)

density

1.292 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc(c1)N=C=S

InChI

1S/C7H4ClNS/c8-6-2-1-3-7(4-6)9-5-10/h1-4H

InChI key

WGXCKFMVBAOIFH-UHFFFAOYSA-N

Application

3-Chlorophenyl isothiocyanate has been used in the synthesis of:
  • 2-[(3-chlorophenyl)amino]naphtho[2,1-b]furo-5H-[3,2-d][1,3,4]thiadiazolo[3,2-i]pyrimidin-5-one
  • (S)-N-[3-{N-(3-chlorophenyl)-4-(3-fluorophenyl)piperazine]-1-carbothioamido}-2-oxooxazolidin-5-yl)methyl]acetamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

253766-BULK:
253766-25G:
253766-5G:
253766-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of novel naphtho [2, 1-b] furo-5H-[3, 2-d][1, 3, 4] thiadiazolo [3, 2-a] pyrimidin-5-ones.
Ravindra KC, et al.
ARKIVOC (Gainesville, FL, United States), 11(1) (2008)
Sunil Kumar Reddy Aaramadaka et al.
Chemical & pharmaceutical bulletin, 55(2), 236-240 (2007-02-03)
Urea and thiourea derivatives of oxazolidinones were synthesized and their inhibitory activity (MIC) was determined on the bacterial strains which includes clinical isolates and quality control organisms. The structure activity relationships were studied and a 3D-QSAR model was built using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service