Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

244287

Sigma-Aldrich

2,2,2-Tribromoethyl dichlorophosphate

97%

Synonym(s):

2,2,2-Tribromoethyl phosphorodichloridate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CBr3CH2OP(O)Cl2
CAS Number:
Molecular Weight:
399.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

bp

90-92 °C/0.4 mmHg (lit.)

mp

34-36 °C (lit.)

SMILES string

ClP(Cl)(=O)OCC(Br)(Br)Br

InChI

1S/C2H2Br3Cl2O2P/c3-2(4,5)1-9-10(6,7)8/h1H2

InChI key

RGBNHRWLNSWPPI-UHFFFAOYSA-N

Application

Reagent for oligonucleotide and oligodeoxynucleotide synthesis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

244287-25G:
244287-BULK:
244287-5G:
244287-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service