241865
Boron carbide
−60-+230 mesh
Synonym(s):
Carbon tetraboride
About This Item
Recommended Products
particle size
−60-+230 mesh
density
2.51 g/mL at 25 °C (lit.)
SMILES string
B1=BB=C=B1
InChI
1S/CB4/c1-2-4-5-3-1
InChI key
NOJMLSPGQSYAIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PRTR
Class I Designated Chemical Substances
JAN Code
241865-100G:
241865-500G:
241865-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service