Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

235814

Sigma-Aldrich

2,4-Dichloro-5-nitrobenzotrifluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)CF3
CAS Number:
Molecular Weight:
260.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

55-57 °C (lit.)

functional group

chloro
fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(c(Cl)cc1Cl)C(F)(F)F

InChI

1S/C7H2Cl2F3NO2/c8-4-2-5(9)6(13(14)15)1-3(4)7(10,11)12/h1-2H

InChI key

VLVNHMVSVDVAOA-UHFFFAOYSA-N

Application

2,4-Dichloro-5-nitrobenzotrifluoride has been used in the synthesis of:
  • 2-substituted 3,7,8-trichlorodibenzo-p-dioxins
  • new substituted 10 H-phenothiazines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

235814-25G:
235814-VAR:
235814-BULK:
235814-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naveen Gautam et al.
Nucleosides, nucleotides & nucleic acids, 29(3), 178-189 (2010-04-22)
This article describes the synthesis of new substituted 10 H-phenothiazines by Smiles rearrangement. These compounds are then used as a base to form ribofuranosides by treating them with a sugar (1-O-acetyl-2,3,5-tri-O-benzoyl-beta-ribofuranose). On oxidation with hydrogen peroxide in glacial acetic acid
M Romkes et al.
Cancer research, 47(19), 5108-5111 (1987-10-01)
The competitive receptor binding affinities of thirteen 2-substituted 3,7,8-trichlorodibenzo-p-dioxins to hepatic cytosol from rat, mouse, guinea pig, and hamster were determined with [3H]-2,3,7,8-tetrachlorodibenzo-p-dioxin as the radioligand. Multiple parameter linear regression analysis of the binding data from the four species gave

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service