Skip to Content
Merck
All Photos(1)

Key Documents

234338

Sigma-Aldrich

4-Amino-5-hydroxy-2,7-naphthalenedisulfonic acid monosodium salt hydrate

96%

Synonym(s):

H Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
Pricing and availability is not currently available.

Assay

96%


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service