Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

227455

Sigma-Aldrich

Methyl 2,2-dimethyl-3-hydroxypropionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CO2CH3
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

177-178 °C/740 mmHg (lit.)

density

1.036 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

COC(=O)C(C)(C)CO

InChI

1S/C6H12O3/c1-6(2,4-7)5(8)9-3/h7H,4H2,1-3H3

InChI key

KJRFTNVYOAGTHK-UHFFFAOYSA-N

General description

Kinetics of decomposition of methyl 2,2-dimethyl-3-hydroxypropionate to methyl isobutyrate and formaldehyde in a static system has been investigated[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

227455-BULK:
227455-25G:
227455-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Rotinov et al.
The journal of physical chemistry. A, 113(15), 3491-3497 (2009-03-25)
Methyl 2,2-dimethyl-3-hydroxypropionate was found to decompose, in a static system, mainly to methyl isobutyrate and formaldehyde. The reaction rates were affected in packed and unpacked clean Pyrex vessels, demonstrating little but significant surface effect. However, in vessels seasoned with allyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service