223700
2,2,5,5-Tetramethyltetrahydrofuran
97%
Synonym(s):
Tetrahydro-2,2,5,5-tetramethylfuran
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.405 (lit.)
bp
112 °C (lit.)
density
0.811 g/mL at 25 °C (lit.)
SMILES string
CC1(C)CCC(C)(C)O1
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
223700-25G:
223700-VAR:
223700-BULK:
223700-5G:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service