Skip to Content
Merck
All Photos(1)

Documents

Safety Information

21160

Sigma-Aldrich

Calcium α-D-heptagluconate dihydrate

≥98.0%

Synonym(s):

α-D-Glucoheptonic acid calcium salt, D-Glycero-D-glucoheptonic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26CaO16 · 2H2O
CAS Number:
Molecular Weight:
526.46
Beilstein:
6104529
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

loss

≤8% loss on drying

SMILES string

[H]O[H].[H]O[H].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C7H14O8.Ca.2H2O/c2*8-1-2(9)3(10)4(11)5(12)6(13)7(14)15;;;/h2*2-6,8-13H,1H2,(H,14,15);;2*1H2/q;;+2;;/p-2/t2*2-,3-,4+,5-,6-;;;/m11.../s1

InChI key

JBMIEBJMNFOKBC-JRLDONDGSA-L

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

21160-1KG-F:
21160-BULK-F:
21160-VAR-F:
21160-50G-F:
21160-250G-F:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service