Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

190667

Sigma-Aldrich

2-Ethoxybenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CH2OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.532 (lit.)

bp

265 °C (lit.)

density

1.074 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1CO

InChI

1S/C9H12O2/c1-2-11-9-6-4-3-5-8(9)7-10/h3-6,10H,2,7H2,1H3

InChI key

ICJVQAHPHKYCNU-UHFFFAOYSA-N

General description

2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

190667-5G:
190667-25G:
190667-BULK:
190667-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith Smith et al.
Organic & biomolecular chemistry, 1(9), 1560-1564 (2003-08-21)
The acetylation of aryl ethers using acetic anhydride in the presence of zeolites under modest conditions in a solvent-free system gave the corresponding para-acetylated products in high yields. The zeolite can be recovered, regenerated and reused to give almost the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service