Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

179884

Sigma-Aldrich

4,5-Dichlorophthalic acid

99%

Synonym(s):

4,5-Dichloro-1,2-benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2-1,2-(CO2H)2
CAS Number:
Molecular Weight:
235.02
Beilstein:
1877706
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

99%

mp

198-200 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(Cl)c(Cl)cc1C(O)=O

InChI

1S/C8H4Cl2O4/c9-5-1-3(7(11)12)4(8(13)14)2-6(5)10/h1-2H,(H,11,12)(H,13,14)

InChI key

FDOQKGWUMUEJLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

This compound has an extremely unconventional infrared spectrum, resembling that of an amino acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

179884-250G:
179884-50G:
179884-1KG:
179884-BULK:
179884-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service