Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

16564

Sigma-Aldrich

Heptadecanenitrile

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CN
CAS Number:
Molecular Weight:
251.45
Beilstein:
1775016
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

solid

SMILES string

CCCCCCCCCCCCCCCCC#N

InChI

1S/C17H33N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h2-16H2,1H3

InChI key

ZXPWFWWSCFIFII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptadecanenitrile gets adsorbed on gold and copper surface from hexane or acetone solution and adsorbed layers are investigated by reflection infrared spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

16564-1G-F:4548173249483
16564-VAR-F:
16564-5G-F:4548173249490
16564-BULK-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of alkanenitriles and alkanedinitriles on gold and copper.
Steiner UB, et al.
Langmuir, 8(11), 2771-2777 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service